Learn More
Biomatik Corporation Rat Histidine Rich Glycoprotein (HRG) ELISA Kit
Supplier:  Biomatik Corporation EKU04807
Encompass_Preferred
Rat Histidine Rich Glycoprotein (HRG) ELISA Kit for detecting HRG in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.117ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q99PS8. Gene ID: 171016. Target Synonyms: HPRG; HRGP; Histidine-Proline Rich Glycoprotein; Thrombophilia Due To Elevated HRG. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2494