Learn More
Biomatik Corporation Human Insulin Like Growth Factor 1 Receptor (IGF1R) ELISA Kit
Supplier:  Biomatik Corporation EKU04972
Encompass_Preferred
Sample Type: Tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Human. Detection Range: 0.312-20ng/mL. Sensitivity: 0.113ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: P08069. Target Synonyms: CD221; IGF1-R; IGFIR; JTK13. Restriction: For Research Use Only.
Catalog No. 50-229-2519