Learn More
Biomatik Corporation Human Interferon Regulatory Factor 1 (IRF1) ELISA Kit
Supplier:  Biomatik Corporation EKU05118
Encompass_Preferred
Sample Type: Tissue homogenates and other biological fluids. Species Reactivity: Human. Detection Range: 0.312-20ng/mL. Sensitivity: 0.122ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: P10914. Target Synonyms: MAR. Restriction: For Research Use Only.
Catalog No. 50-229-2540