Learn More
Biomatik Corporation Guinea pig Interleukin 1 Alpha (IL1a) ELISA Kit
Supplier:  Biomatik Corporation EKU05129
Encompass_Preferred
Guinea pig Interleukin 1 Alpha (IL1a) ELISA Kit for detecting IL1a in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Guinea pig (Cavia porcellus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 5.5pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q60480. Target Synonyms: IL1-A; IL-1α; IL1F1; Preinterleukin 1 Alpha; Hematopoietin-1; Pro-Interleukin-1-Alpha; Interleukin-1 Family Member 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2543