Learn More
Biomatik Corporation Horse Interleukin 1 Receptor Antagonist (IL1RA) ELISA Kit
Supplier:  Biomatik Corporation EKU05155
Encompass_Preferred
Horse Interleukin 1 Receptor Antagonist (IL1RA) ELISA Kit for detecting IL1RA in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Horse (Equus caballus; Equine). Detection Range: 15.6-1,000pg/ml. Sensitivity: 6.2pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O18999. Gene ID: 100034236. Target Synonyms: IL1RN; IL1-RA; ICIL1RA; IL1ra3; IL1F3; IRAP;IL-1RA; Interleukin-1 Family Member 3; IL1 inhibitor; Anakinra. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2547