Learn More
Biomatik Corporation Goat Interleukin 10 (IL10) ELISA Kit
Supplier:  Biomatik Corporation EKU05174
Encompass_Preferred
Goat Interleukin 10 (IL10) ELISA Kit for detecting IL10 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Goat (Capra aegagrus hircus). Detection Range: 3.12-200pg/ml. Sensitivity: 1.18pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: A7Y3X5. Target Synonyms: CSIF; IL10A; TGIF; Cytokine Synthesis Inhibitory Factor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2552