Learn More
Biomatik Corporation Human Interleukin-18 Receptor Beta (IL18R Beta) ELISA Kit
Supplier:  Biomatik Corporation EKU05262
Encompass_Preferred
Sample Type: Tissue homogenates and other biological fluids.. Species Reactivity: Human. Detection Range: 0.156-10ng/mL. Sensitivity: 0.051ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: Q3KPE8. Target Synonyms: CDw218b; IL18RAP; CD218-B; IL-1R7; ACPL; Interleukin 18 Receptor Accessory Protein; Accessory protein-likeL; CD218 antigen-like family member B; IL-1R accessory protein-like; Interleukin-1 receptor 7;. Restriction: For Research Use Only.
Catalog No. 50-229-2562