Learn More
Biomatik Corporation Mouse Interleukin 2 Receptor Alpha (IL2Ra) ELISA Kit
Supplier:  Biomatik Corporation EKU05279
Encompass_Preferred
Mouse Interleukin 2 Receptor Alpha (IL2Ra) ELISA Kit for detecting IL2Ra in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 6.2pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P01590. Gene ID: 16184. Target Synonyms: CD25; IL2R-a; IL2-RA; IDDM10; IL2R; TCGFR; p55; TAC antigen. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2564