Learn More
Biomatik Corporation Human Interleukin 28A (IL28A) ELISA Kit
Supplier:  Biomatik Corporation EKU05309
Encompass_Preferred
Human Interleukin 28A (IL28A) ELISA Kit for detecting IL28A in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.78-50ng/ml. Sensitivity: 0.31ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q8IZJ0. Gene ID: 282616. Target Synonyms: IFNL2; Interferon,Lambda 2; Cytokine Zcyto20. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2567