Learn More
Biomatik Corporation Mouse Interleukin 7 (IL7) ELISA Kit
Supplier:  Biomatik Corporation EKU05370
Encompass_Preferred
Mouse Interleukin 7 (IL7) ELISA Kit for detecting IL7 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.6-1000pg/ml. Sensitivity: 6.2pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P10168. Gene ID: 16196. Target Synonyms: Pre-B-cell Growth Factor; Lymphopoietin-1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2575