Learn More
Biomatik Corporation Simian Interleukin 8 (IL8) ELISA Kit
Supplier:  Biomatik Corporation EKU05382
Encompass_Preferred
Simian Interleukin 8 (IL8) ELISA Kit for detecting IL8 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Simian. Detection Range: 15.6-1,000pg/ml. Sensitivity: 5.7pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P67813. Gene ID: 613028. Target Synonyms: CXCL8; AMCF-I; GCP1; K60; LECT; LUCT; LYNAP; MDNCF; MONAP; NAF; NAP1; SCYB8; TSG1; B-ENAP; Neutrophil-Activating Protein 1; Granulocyte Chemotactic Protein 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2578