Learn More
Biomatik Corporation Human Kallistatin (KAL) ELISA Kit
Supplier:  Biomatik Corporation EKU05455
Encompass_Preferred
Human Kallistatin (KAL) ELISA Kit for detecting KAL in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 6.25-400ng/ml. Sensitivity: 2.95ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P29622. Gene ID: 5267. Target Synonyms: SERPINA4; KLST; KST; PI4; Serpin Peptidase Inhibitor Clade A Member 4; Tissue Kallikrein Inhibitor; Kallikrein Binding Protein; Peptidase inhibitor 4. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2590