Learn More
Biomatik Corporation Human Keratin 2 (CK2) ELISA Kit
Supplier:  Biomatik Corporation EKU05472
Encompass_Preferred
Human Keratin 2 (CK2) ELISA Kit for detecting CK2 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.059ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P35908. Gene ID: 3849. Target Synonyms: KRT2A; KRT2; KRTE; Keratin 2; Keratin 2A; Keratin 2E; Epidermal Ichthyosis Bullosa Of Siemens; Keratin, type II cytoskeletal 2 epidermal; Epithelial keratin-2e. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2592