Learn More
Biomatik Corporation Human Legumain (LGMN) ELISA Kit
Supplier:  Biomatik Corporation EKU05585
Encompass_Preferred
Human Legumain (LGMN) ELISA Kit for detecting LGMN in serum, plasma, tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.123ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q99538. Gene ID: 5641. Target Synonyms: AEP; LGMN1; PRSC1; Protease,Cysteine 1; Asparaginyl endopeptidase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2610