Learn More
Biomatik Corporation Mouse Leptin Receptor (LEPR) ELISA Kit
Supplier:  Biomatik Corporation EKU05599
Encompass_Preferred
Mouse Leptin Receptor (LEPR) ELISA Kit for detecting LEPR in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.063ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P48356. Gene ID: 16847. Target Synonyms: CD295; LEPR; OBR; LR. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2613