Learn More
Biomatik Corporation Mouse Leukocyte Elastase Inhibitor (LEI) ELISA Kit
Supplier:  Biomatik Corporation EKU05626
Encompass_Preferred
Mouse Leukocyte Elastase Inhibitor (LEI) ELISA Kit for detecting LEI in serum, plasma, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 6.7pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9D154. Gene ID: 66222. Target Synonyms: SERPINB1; EI; ELANH2; LEI; M/NEI; MNEI; PI2; Serpin Peptidase Inhibitor Clade B Member 1; Anti-Elastase; Monocyte/Neutrophil Elastase Inhibitor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2616