Learn More
Biomatik Corporation Mouse Lipase, Endothelial (LIPG) ELISA Kit
Supplier:  Biomatik Corporation EKU05641
Encompass_Preferred
Mouse Lipase, Endothelial (LIPG) ELISA Kit for detecting LIPG in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 1.56-100ng/ml. Sensitivity: 0.60ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9WVG5. Gene ID: 16891. Target Synonyms: EL; EDL; Endothelial Lipase; Endothelial cell-derived lipase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2618