Learn More
Biomatik Corporation Mouse Lipase, Hepatic (LIPC) ELISA Kit
Supplier:  Biomatik Corporation EKU05645
Encompass_Preferred
Mouse Lipase, Hepatic (LIPC) ELISA Kit for detecting LIPC in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 3.12-200ng/ml. Sensitivity: 1.13ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P27656. Gene ID: 15450. Target Synonyms: HL; HTGL; Hepatic Lipase; Hepatic triacylglycerol lipase; Lipase member C. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2620