Learn More
Biomatik Corporation Human Low Density Lipoprotein Receptor Related Protein 4 (LRP4) ELISA Kit
Supplier:  Biomatik Corporation EKU05683
Encompass_Preferred
Sample Type: Tissue homogenates, cell lysates and other biological fluids.. Species Reactivity: Human. Detection Range: 0.312-20ng/mL. Sensitivity: 0.126ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: O75096. Target Synonyms: MEGF7; LRP10; Multiple epidermal growth factor-like domains 7. Restriction: For Research Use Only.
Catalog No. 50-229-2623