Learn More
Biomatik Corporation Human Low Molecular Weight Kininogen (LMWK) ELISA Kit
Supplier:  Biomatik Corporation EKU05687
Encompass_Preferred
Human Low Molecular Weight Kininogen (LMWK) ELISA Kit for detecting LMWK in plasma, tissue homogenates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 6.25-400ng/ml. Sensitivity: 2.55ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P01042. Gene ID: 3827. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2625