Learn More
Biomatik Corporation Human Lymphotoxin Beta Receptor (LTbR) ELISA Kit
Supplier:  Biomatik Corporation EKU05709
Encompass_Preferred
Sample Type: Tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Human. Detection Range: 0.312-20ng/mL. Sensitivity: 0.115ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: P36941. Target Synonyms: TNFCR; TNFR-RP; TNFR-III; TNFR2-RP; TNFRSF3; Tumor Necrosis Factor Superfamily Member 3; Tumor necrosis factor C receptor; Tumor necrosis factor receptor type III. Restriction: For Research Use Only.
Catalog No. 50-229-2629