Learn More
Biomatik Corporation Human Mannose Receptor C Type 1 Like Protein 1 (MRC1) ELISA Kit
Supplier:  Biomatik Corporation EKU05796
Encompass_Preferred
Sample Type: Tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Human. Detection Range: 0.156-10ng/mL. Sensitivity: 0.054ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: Not Available. Target Synonyms: CD206; CLEC13DL; MRC1L1; C-Type Lectin Domain Family 13 Member D-Like; Macrophage mannose receptor 1; Macrophage mannose receptor 1-like protein 1. Restriction: For Research Use Only.
Catalog No. 50-229-2642