Learn More
Biomatik Corporation Human Mammary Serine Protease Inhibitor (Maspin) ELISA Kit
Supplier:  Biomatik Corporation EKU05804
Encompass_Preferred
Human Mammary Serine Protease Inhibitor (Maspin) ELISA Kit for detecting Maspin in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.625-40ng/ml. Sensitivity: 0.233ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P36952. Gene ID: 5268. Target Synonyms: SERPINB5; PI5; Protease Inhibitor 5; Serpin Peptidase Inhibitor Clade B Member 5. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2645