Learn More
Biomatik Corporation Rat Matrix Gla Protein (MGP) ELISA Kit
Supplier:  Biomatik Corporation EKU05815
Encompass_Preferred
Rat Matrix Gla Protein (MGP) ELISA Kit for detecting MGP in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.78-50ng/ml. Sensitivity: 0.29ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P08494. Gene ID: 25333. Target Synonyms: MGLAP; GIG36; Cell growth-inhibiting gene 36 protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2647