Learn More
Biomatik Corporation Human Matrix Metalloproteinase 17 (MMP17) ELISA Kit
Supplier:  Biomatik Corporation EKU05846
Encompass_Preferred
Human Matrix Metalloproteinase 17 (MMP17) ELISA Kit for detecting MMP17 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.056ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9ULZ9. Gene ID: 4326. Target Synonyms: MT4-MMP; MMP4; Membrane-type matrix metalloproteinase 4; Membrane-type-4 matrix metalloproteinase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2654