Learn More
Biomatik Corporation Mouse Matrix Metalloproteinase 8 (MMP8) ELISA Kit
Supplier:  Biomatik Corporation EKU05876
Encompass_Preferred
Mouse Matrix Metalloproteinase 8 (MMP8) ELISA Kit for detecting MMP8 in serum, plasma, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 31.2-2,000pg/ml. Sensitivity: 12.7pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O70138. Gene ID: 17394. Target Synonyms: CLG2; HNC; PMNL-CL; Collagenase 2; Neutrophil Collagenase; PMNL collagenase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2664