Learn More
Biomatik Corporation Mouse Metallothionein 2 (MT2) ELISA Kit
Supplier:  Biomatik Corporation EKU05922
Encompass_Preferred
Mouse Metallothionein 2 (MT2) ELISA Kit for detecting MT2 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 31.2-2,000pg/ml. Sensitivity: 12.1pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P02798. Gene ID: 17750. Target Synonyms: MT2A; CES1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2667