Learn More
Biomatik Corporation Human Methionine Adenosyltransferase II Alpha (MAT2a) ELISA Kit
Supplier:  Biomatik Corporation EKU05933
Encompass_Preferred
Sample Type: Tissue homogenates, cell lysates and other biological fluids.. Species Reactivity: Human. Detection Range: 0.156-10ng/mL. Sensitivity: 0.054ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: P31153. Target Synonyms: SAMS2; MATA2; AMS2; MATII; S-adenosylmethionine synthase isoform type-2. Restriction: For Research Use Only.
Catalog No. 50-229-2670