Learn More
Biomatik Corporation Mouse Microsomal Triglyceride Transfer Protein (MTTP) ELISA Kit
Supplier:  Biomatik Corporation EKU05947
Encompass_Preferred
Sample Type: tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Mouse. Detection Range: 0.156-10ng/mL. Sensitivity: 0.056ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: O08601. Target Synonyms: MTP; ABL; Microsomal triglyceride transfer protein large subunit. Restriction: For Research Use Only.
Catalog No. 50-229-2673