Learn More
Biomatik Corporation Mouse Milk Fat Globule EGF Factor 8 (MFGE8) ELISA Kit
Supplier:  Biomatik Corporation EKU05964
Encompass_Preferred
Mouse Milk Fat Globule EGF Factor 8 (MFGE8) ELISA Kit for detecting MFGE8 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 5.8pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P21956. Gene ID: 17304. Target Synonyms: HMFG; MFGM; SED1; BA46; EDIL1; OAcGD3S; SPAG10; hP47; Medin; Lactadherin; Sperm Associated Antigen 10; Sperm Surface Protein hP47; Milk fat globule-EGF factor 8. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2678