Learn More
Biomatik Corporation Human Mucin 13, Cell Surface Associated (MUC13) ELISA Kit
Supplier:  Biomatik Corporation EKU06018
Encompass_Preferred
Sample Type: Tissue homogenates, cell lysates, cell culture supernates and other biological fluids. Species Reactivity: Human. Detection Range: 0.156-10ng/mL. Sensitivity: 0.059ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: Q9H3R2. Target Synonyms: DRCC1; RECC; Down-Regulated In Colon Cancer 1. Restriction: For Research Use Only.
Catalog No. 50-229-2686