Learn More
Biomatik Corporation Human Myomesin 2 (MYOM2) ELISA Kit
Supplier:  Biomatik Corporation EKU06083
Encompass_Preferred
Sample Type: tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Human. Detection Range: 0.312-20ng/mL. Sensitivity: 0.113ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: P54296. Target Synonyms: TTNAP; Myomesin family member 2; 165 kDa titin-associated protein; 165 kDa connectin-associated protein. Restriction: For Research Use Only.
Catalog No. 50-229-2692