Learn More
Biomatik Corporation Human Myosin Heavy Chain 3, Skeletal Muscle, Embryonic (MYH3) ELISA Kit
Supplier:  Biomatik Corporation EKU06093
Encompass_Preferred
Sample Type: tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Human. Detection Range: 0.312-20ng/mL. Sensitivity: 0.121ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: P11055. Target Synonyms: HEMHC; MYHC-EMB; MYHSE1; SMHCE; Myosin,Skeletal,Heavy chain,Embryonic 1; Myosin heavy chain, fast skeletal muscle, embryonic. Restriction: For Research Use Only.
Catalog No. 50-229-2694