Learn More
Biomatik Corporation Mouse Myosin Heavy Chain 7, Cardiac Muscle, Beta (MYH7) ELISA Kit
Supplier:  Biomatik Corporation EKU06099
Encompass_Preferred
Mouse Myosin Heavy Chain 7, Cardiac Muscle, Beta (MYH7) ELISA Kit for detecting MYH7 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 31.2-2,000pg/ml. Sensitivity: 11.2pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q91Z83. Gene ID: 140781. Target Synonyms: CMD1S; CMH1; MPD1; MYHCB; Myopathy,Distal 1; Myosin heavy chain slow isoform; Myosin heavy chain, cardiac muscle beta isoform. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2696