Learn More
Biomatik Corporation Pig Myostatin (MSTN) ELISA Kit
Supplier:  Biomatik Corporation EKU06117
Encompass_Preferred
Pig Myostatin (MSTN) ELISA Kit for detecting MSTN in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Pig (Sus scrofa; Porcine). Detection Range: 125-8,000pg/ml. Sensitivity: 53pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O18831. Target Synonyms: GDF8; Growth Differentiation Factor 8. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2699