Learn More
Biomatik Corporation Rat NADH Dehydrogenase, Quinone 1 (NQO1) ELISA Kit
Supplier:  Biomatik Corporation EKU06149
Encompass_Preferred
Sample Type: Tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Rat. Detection Range: 0.312-20ng/mL. Sensitivity: 0.114ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: P05982. Target Synonyms: DHQU; DIA4; DTD; NMOR1; NMORI; QR1; Azoreductase; DT-diaphorase; Menadione reductase; Phylloquinone reductase; Quinone reductase 1; NAD(P)H:quinone oxidoreductase 1. Restriction: For Research Use Only.
Catalog No. 50-229-2706