Learn More
Biomatik Corporation Bovine Neuropeptide Y (NPY) ELISA Kit
Supplier:  Biomatik Corporation EKU06246
Encompass_Preferred
Bovine Neuropeptide Y (NPY) ELISA Kit for detecting NPY in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Bovine (Bos taurus). Detection Range: 24.69-2000pg/ml. Sensitivity: 9.35pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: Q6RUW3. Gene ID: 504216. Target Synonyms: NP-Y; PYY4; CPON; Pro-neuropeptide Y; Neuropeptide tyrosine; C-flanking peptide of NPY. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2721