Learn More
Biomatik Corporation Rat Neurotrophin 3 (NT3) ELISA Kit
Supplier:  Biomatik Corporation EKU06272
Encompass_Preferred
Rat Neurotrophin 3 (NT3) ELISA Kit for detecting NT3 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 3.12-200pg/ml. Sensitivity: 1.15pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P18280. Gene ID: 81737. Target Synonyms: NTF3; HDNF; NGF2; Nerve Growth Factor 2; Neurotrophic Factor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2723