Learn More
Biomatik Corporation Human Neutrophil Cytosolic Factor 1 (NCF1) ELISA Kit
Supplier:  Biomatik Corporation EKU06283
Encompass_Preferred
Sample Type: tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Human. Detection Range: 0.312-20ng/mL. Sensitivity: 0.115ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: P14598. Target Synonyms: NCF1A; NOXO2; SH3PXD1A; p47-phox; NCF-47K; 47 kDa autosomal chronic granulomatous disease protein; Neutrophil NADPH oxidase factor 1; Nox-organizing protein 2. Restriction: For Research Use Only.
Catalog No. 50-229-2726