Learn More
Biomatik Corporation Human Nidogen (NID) ELISA Kit
Supplier:  Biomatik Corporation EKU06299
Encompass_Preferred
Human Nidogen (NID) ELISA Kit for detecting NID in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.059ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P14543. Gene ID: 4811. Target Synonyms: Entactin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2728