Learn More
Biomatik Corporation Mouse Octamer Binding Transcription Factor 4 (OCT4) ELISA Kit
Supplier:  Biomatik Corporation EKU06385
Encompass_Preferred
Sample Type: tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Mouse. Detection Range: 0.156-10ng/mL. Sensitivity: 0.055ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: P20263. Target Synonyms: POU5F1; OCT3; OTF3; OTF4; Octamer-4; POU Domain Class 5 Transcription Factor 1; POU Class 5 Homoeobox 1; Octamer-binding protein 3. Restriction: For Research Use Only.
Catalog No. 50-229-2738