Learn More
Biomatik Corporation Mouse Optineurin (OPTN) ELISA Kit
Supplier:  Biomatik Corporation EKU06395
Encompass_Preferred
Sample Type: Tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Mouse. Detection Range: 0.156-10ng/mL. Sensitivity: 0.057ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: Q8K3K8. Target Synonyms: NRP; FIP2; GLC1E; HIP7; HYPL; TFIIIA-INTP; Glaucoma 1,Open Angle,E(Adult-Onset); Huntingtin-interacting protein 7; Optic neuropathy-inducing protein; E3-14.7K-interacting protein. Restriction: For Research Use Only.
Catalog No. 50-229-2741