Learn More
Biomatik Corporation Rat Osteocrin (OSTN) ELISA Kit
Supplier:  Biomatik Corporation EKU06421
Encompass_Preferred
Rat Osteocrin (OSTN) ELISA Kit for detecting OSTN in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.058ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P61365. Gene ID: 360730. Target Synonyms: Musclin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2743