Learn More
Biomatik Corporation Mouse Oxoguanine Glycosylase 1 (OGG1) ELISA Kit
Supplier:  Biomatik Corporation EKU06451
Encompass_Preferred
Sample Type: tissue homogenates, cell lysates, cell culture supernates and other biological fluids. Species Reactivity: Mouse. Detection Range: 31.25-2,000pg/mL. Sensitivity: 12.5pg/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: O08760. Target Synonyms: HMMH; HOGG1; MUTM; OGH1; 8-Oxoguanine DNA Glycosylase; DNA-(apurinic or apyrimidinic site) lyase; N-glycosylase/DNA lyase. Restriction: For Research Use Only.
Catalog No. 50-229-2747