Learn More
Biomatik Corporation Human Phosphodiesterase 4D, cAMP Specific (PDE4D) ELISA Kit
Supplier:  Biomatik Corporation EKU06592
Encompass_Preferred
Sample Type: tissue homogenates, cell lysates, cell culture supernates and other biological fluids. Species Reactivity: Human. Detection Range: 0.312-20ng/mL. Sensitivity: 0.113ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: Q08499. Target Synonyms: DPDE3; PDE43; Dunce(Drosophila)Homolog Phosphodiesterase E3; cAMP-specific 3',5'-cyclic phosphodiesterase 4D. Restriction: For Research Use Only.
Catalog No. 50-229-2763