Learn More
Biomatik Corporation Human Phospholipase C Gamma 2, Phosphatidylinositol Specific (PLCg2) ELISA Kit
Supplier:  Biomatik Corporation EKU06628
Encompass_Preferred
Sample Type: Tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Human. Detection Range: 0.312-20ng/mL. Sensitivity: 0.125ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: P16885. Target Synonyms: PLC-IV; Phosphoinositide phospholipase C-gamma-2; 1-phosphatidylinositol 4,5-bisphosphate phosphodiesterase gamma-2. Restriction: For Research Use Only.
Catalog No. 50-229-2770