Learn More
Biomatik Corporation Human Platelet Derived Growth Factor Receptor Like Protein (PDGFRL) ELISA Kit
Supplier:  Biomatik Corporation EKU06694
Encompass_Preferred
Human Platelet Derived Growth Factor Receptor Like Protein (PDGFRL) ELISA Kit for detecting PDGFRL in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 3.12-200ng/ml. Sensitivity: 1.13ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q15198. Gene ID: 5157. Target Synonyms: PRLTS; PDGF-Receptor Beta-Like Tumor Suppressor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2779