Learn More
Biomatik Corporation Human Polymerase DNA Directed Gamma 1 (POLg1) ELISA Kit
Supplier:  Biomatik Corporation EKU06740
Encompass_Preferred
Sample Type: Tissue homogenates, cell lysates, cell culture supernates and other biological fluids. Species Reactivity: Human. Detection Range: 0.312-20ng/mL. Sensitivity: 0.113ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: P54098. Target Synonyms: PEO; POLG1; POLGA; SANDO; SCAE; Mitochondrial DNA polymerase catalytic subunit. Restriction: For Research Use Only.
Catalog No. 50-229-2783