Learn More
Biomatik Corporation Human 17-Beta-Hydroxysteroid Dehydrogenase Type 3 (HSD17b3) ELISA Kit
Supplier:  Biomatik Corporation EKU02010
Encompass_Preferred
Sample Type: Tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Human. Detection Range: 0.312-20ng/mL. Sensitivity: 0.113ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: P37058. Target Synonyms: SDR12C2; EDH17B3; Short Chain Dehydrogenase/Reductase Family 12C,Member 2; Testosterone 17-Beta-Dehydrogenase 3; Testicular 17-beta-hydroxysteroid dehydrogenase. Restriction: For Research Use Only.
Catalog No. 50-229-2042