Learn More
Biomatik Corporation Rat A Disintegrin And Metalloproteinase With Thrombospondin 4 (ADAMTS4) ELISA Kit
Supplier:  Biomatik Corporation EKU02058
Encompass_Preferred
Rat A Disintegrin And Metalloproteinase With Thrombospondin 4 (ADAMTS4) ELISA Kit for detecting ADAMTS4 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.056ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: D4A0C5. Target Synonyms: ADMP-1; Aggrecanase-1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-229-2049